missing translation for 'onlineSavingsMsg'
Learn More

Sodium thiosulfate, 0.01N Standardized Solution

Catalog No. AA35643K2
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA35643K2 1 L
AA35643K7 4 L
AA35643M4 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA35643K2 Supplier Thermo Scientific Chemicals Supplier No. 035643K2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7772-98-7 | Na2O3S2 | 158.10 g/mol

Sodium thiosulfate, 0.01N standardized solution is used in volumetric analysis to estimate the concentration of certain compounds in solution (hydrogen peroxide) as well as chlorine content in bleaching powder and water. It is used as a titrant in iodometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7772-98-7
Molecular Formula Na2O3S2
Molecular Weight (g/mol) 158.10
MDL Number MFCD00003499
InChI Key AKHNMLFCWUSKQB-UHFFFAOYSA-L
Synonym sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt
PubChem CID 24477
IUPAC Name disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane
SMILES [Na+].[Na+].[O-]S([S-])(=O)=O

Specifications

Concentration 0.01N
Linear Formula Na2S2O3
Quantity 1 L
Physical Form Liquid
Solubility Information Immiscible with alcohol.
Formula Weight 158.1
Chemical Name or Material Sodium thiosulfate, Standardized Solution
EINECSNumber 231-867-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only