accessibility menu, dialog, popup

UserName

KKOOOOH2O

CAS RN 6487-48-5

IUPAC Name: dipotassium hydrate oxalate
Synonyms: potassium oxalate monohydrate
Molecular Weight (g/mol): 184.23
Molecular Formula: C2H2K2O5
InChi Key: QCPTVXCMROGZOL-UHFFFAOYSA-L
SMILES: O.[K+].[K+].[O-]C(=O)C([O-])=O
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (5)
  • (2)
  • (1)
  • (8)
  • (1)
Grade 
  • (7)
  • (2)
  • (5)
  • (2)
  • (1)
  • (5)
Physical Form 
  • (2)
  • (4)
  • (5)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (7)
  • (3)
  • (5)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (11)
  • (9)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (9)
  • (10)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (5)
  • (2)
  • (1)
  • (8)
  • (1)
  • (7)
  • (2)
  • (5)
  • (2)
  • (1)
  • (5)
  • (2)
  • (4)
  • (5)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (7)
  • (3)
  • (5)
  • (2)