missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Oxalate, Monohydrate, CP, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Physical Form | Crystalline Powder or Needles |
| Quantity | 100 lb. |
| Molecular Weight (g/mol) | 184.23 |
| Grade | Not Available |
| Packaging | Poly Bottle |
| Chemical Name or Material | Potassium Oxalate, Monohydrate |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.