missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Oxalate, Monohydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Specifications
Specifications
| Physical Form | Crystals |
| Quantity | 2.5 kg |
| Formula Weight | 184.23 |
| Percent Purity | 98.5 to 101.0% |
| Grade | BAKER ANALYZED™ ACS Reagent |
| Packaging | Wide Mouth Amber Glass Bottle |
| Specific Gravity | 1L = 2.13kg |
| Chemical Name or Material | Potassium Oxalate, Monohydrate |