missing translation for 'onlineSavingsMsg'
Learn More

Potassium Oxalate Monohydrate, ACS Reagent Grade, Ricca Chemical

Catalog No. RDCP0610500
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDCP0610500 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RDCP0610500 Supplier Ricca Chemical Company Supplier No. RDCP0610500B1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6487-48-5
Molecular Formula C2H2K2O5
Molecular Weight (g/mol) 184.23
InChI Key QCPTVXCMROGZOL-UHFFFAOYSA-L
Synonym potassium oxalate monohydrate, potassium oxalate hydrate, oxalic acid potassium salt, ethanedioic acid, dipotassium salt, monohydrate, dipotassium oxalate hydrate, dipotassium hydrate oxalate, acmc-1bcm6, ksc495a4h, dipotassium oxalate monohydrate, dipotassium ethanedioate hydrate
PubChem CID 2724193
IUPAC Name dipotassium;oxalate;hydrate
SMILES C(=O)(C(=O)[O-])[O-].O.[K+].[K+]

Specifications

CAS Min % 98.0
CAS Max % 100.0
Quantity 500 g
Grade ACS
Packaging White Poly Wide Mouth Bottle
Chemical Name or Material Potassium Oxalate, Monohydrate
ShelfLife 60 months