missing translation for 'onlineSavingsMsg'
Learn More

Potassium Oxalate, Monohydrate, Crystal, ACS, 98.5-101.0%, Spectrum™ Chemical

Catalog No. 18612819
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612819 50 kg Poly Bottle
18612815 125 g Poly Bottle
18612816 500 g Poly Bottle
18612817 2.5 kg Poly Bottle
18612818 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18612819 Supplier Spectrum Chemical Mfg Cor Supplier No. P135550KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6487-48-5

Specifications

CAS Max % 1
Physical Form Crystalline Powder, Needles or Crystals
Quantity 50 kg
Molecular Weight (g/mol) 184.23
Percent Purity 98.5 to 101.0%
Grade ACS/Reagent
Packaging Poly Bottle
Chemical Name or Material Potassium Oxalate, Monohydrate, Crystal

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.