missing translation for 'onlineSavingsMsg'
Learn More

Potassium Oxalate, Monohydrate, ACS Grade, 98.5% to 101.0%, LabChem™

Catalog No. LC198351
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC198351 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC198351 Supplier LabChem Supplier No. LC198351

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6487-48-5
Molecular Formula C2H2K2O5
Molecular Weight (g/mol) 184.23
MDL Number MFCD00150033
InChI Key QCPTVXCMROGZOL-UHFFFAOYSA-L
Synonym potassium oxalate monohydrate, potassium oxalate hydrate, oxalic acid potassium salt, ethanedioic acid, dipotassium salt, monohydrate, dipotassium oxalate hydrate, dipotassium hydrate oxalate, acmc-1bcm6, ksc495a4h, dipotassium oxalate monohydrate, dipotassium ethanedioate hydrate
PubChem CID 2724193
IUPAC Name dipotassium hydrate oxalate
SMILES O.[K+].[K+].[O-]C(=O)C([O-])=O

Specifications

Color White
CAS Max % 100
Physical Form Crystals
Quantity 500 g
Linear Formula K2C2O4·H2O
Solubility Information Soluble in water
Formula Weight 184.24
Percent Purity 98.5 to 101.0%
Grade ACS
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 2130kg/m3
Density 2,130kg/m3
Chemical Name or Material Potassium Oxalate, Monohydrate
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Harmful if swallowed or in contact with skin.
Precautionary Statement GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
If swallowed: Rinse mouth.
Call a poison center or doctor if you feel unwell.
If on skin: Wash with plenty of soap and water.
Call a poison center/doctor if you feel unwell.
Take off contaminated clothing and wash before reuse.
Dispose of contents/container to comply with local, state and federal regulations.
EINECSNumber 209-506-8
Recommended Storage Room Temperature