CAS RN 63181-94-2
CAS RN 63181-94-2
IUPAC Nor:
1,2-diethenylbenzene [(2-ethenylphenyl)methyl](2-hydroxyethyl)dimethylazanium chloride
Synonymes:
[(2-ethenylphenyl)methyl](2-hydroxyethyl)dimethylazanium divinylbenzene chloride
Poids moléculaire (g/mol):
371.95
Formule moléculaire:
C23H30ClNO
InChi Key:
DPRMXNMXCFHCAS-UHFFFAOYSA-M
SMILES:
[Cl-].C=CC1=CC=CC=C1C=C.C[N+](C)(CCO)CC1=CC=CC=C1C=C
1
–
2
de
2
résultats
1
2
MilliporeSigma™ Supelco™ Amberlite™ IRA-410 Chloride Form, Matrix styrene/divinylbenzene, 20-25 mesh
Strongly basic (type II) anion exchange resin for water conditioning, removing iodine ions, neutralization of solutions, capturing xanthan and xanthene dyes, etc.; better regeneration efficiency than type I base resin.