missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Amberlite™ IRA-410 Chloride Form, Matrix styrene/divinylbenzene (gel), 20-25 mesh

Catalog No. 111014978
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014978 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014978 Supplier MilliporeSigma™ Supelco™ Supplier No. 064331KG

Strongly basic

Amberlite™ IRA-410, anion exchange resin, is an effective catalyst especially for those reactions involving low catalyst concentrations and relatively low temperature. Amberlite™ IRA-410 is a gel-type resin having styrene-divinyl-benzene skeletal structure.

Specifications

Particle Size 20/25 Mesh
pH 0 to 14
Quantity 1 kg
Product Line Amberlite
Capacity (Metric) 1.25 meq/mL by wetted bed volume
MDL Number MFCD00132710
CAS 63181-94-2

Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow