accessibility menu, dialog, popup

UserName

(S)NH2OOH

CAS RN 63-91-2

IUPAC Name: 2-amino-3-phenylpropanoic acid
Synonyms: phenylalanin sabiden phenylalanine DL-form DL-3-phenylalanine alanine, phenyl-, DL- (8CI) alanine, phenyl-, D- alanine, phenyl-, dl- fenilalanina
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: COLNVLDHVKWLRT-UHFFFAOYNA-N
SMILES: NC(CC1=CC=CC=C1)C(O)=O
Quantity 
  • (2)
  • (7)
  • (1)
  • (1)
  • (8)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (5)
  • (3)
  • (5)
  • (1)
Grade 
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (5)
Color 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (19)
  • (8)
  • (2)
  • (1)
  • (2)
  • (8)
  • (2)
  • (14)
  • (2)
  • (7)
  • (1)
  • (1)
  • (8)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (5)
  • (3)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (5)
  • (1)