missing translation for 'onlineSavingsMsg'
Learn More

L-Phenylalanine 98.0+%, TCI America™

Catalog No. P013425G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P013425G 25 g
P0134250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P013425G Supplier TCI America Supplier No. P013425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 63-91-2 | C9H11NO2 | 165.19 g/mol

Chemical Identifiers

CAS 63-91-2
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00064227
InChI Key COLNVLDHVKWLRT-UHFFFAOYNA-N
Synonym l-phenylalanine, phenylalanine, s-2-amino-3-phenylpropanoic acid, 3-phenyl-l-alanine, s-phenylalanine, 3-phenylalanine, 2s-2-amino-3-phenylpropanoic acid, s-2-amino-3-phenylpropionic acid, beta-phenyl-l-alanine, h-phe-oh
PubChem CID 6140
ChEBI CHEBI:17295
IUPAC Name 2-amino-3-phenylpropanoic acid
SMILES NC(CC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 275°C
Color White
Quantity 25 g
Formula Weight 165.19
Percent Purity ≥98.0% (HPLC,T)
Grade Research
Physical Form Crystalline Powder
Chemical Name or Material L-Phenylalanine
EINECSNumber (9)-1283
RTECSNumber AY7535000
TSCA Yes