missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-phenylalanine, 99.85%, MP Biomedicals™
Spécifications
Spécifications
| Color | White |
| Quantity | 25 g |
| Formula Weight | 165.2 |
| Percent Purity | 99.85% |
| Grade | Biochemical |
| Physical Form | Powder |
| Chemical Name or Material | L-Phenylalanine |