missing translation for 'onlineSavingsMsg'
Learn More

Phenylalanine, USP, 98.5-101.5%, Spectrum™ Chemical

Catalog No. 18605125
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605125 50 kg Fiber Drum
18605121 25 g Poly Bottle
18605122 100 g Poly Bottle
18605123 1 kg Poly Pail
18605124 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18605125 Supplier Spectrum Chemical Mfg Cor Supplier No. P158250KGBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

P1582, 63-91-2, C9H11NO2

Spectrum™ Chemical Phenylalanine, USP is an amino acid, a "building block" of protein. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 63-91-2
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
InChI Key COLNVLDHVKWLRT-UHFFFAOYNA-N
IUPAC Name 2-amino-3-phenylpropanoic acid
SMILES NC(CC1=CC=CC=C1)C(O)=O

Specifications

CAS Max % 1
pH 5.4 to 6.0
Chloride 0.0005
Ignition Residue 0.004
Iron (Fe) 3e-005
Loss on Drying 0.003
Packaging Fiber Drum
Specific Rotation -32.7° to -34.7°
Sulfate 0.0003
Quantity 50 kg
Percent Purity 98.5 to 101.5%
Grade USP
Show More Show Less