missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ L-Phenylalanine, Calbiochem™,

Catalog No. 5202100GM
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5202100GM 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5202100GM Supplier MilliporeSigma™ Supplier No. 5202100GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Essential amino acid for diagnostic assay manufacturing, hematology, and histology applications.

Chemical Identifiers

Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
InChI Key COLNVLDHVKWLRT-UHFFFAOYNA-N
Synonym l-phenylalanine, phenylalanine, s-2-amino-3-phenylpropanoic acid, 3-phenyl-l-alanine, s-phenylalanine, 3-phenylalanine, 2s-2-amino-3-phenylpropanoic acid, s-2-amino-3-phenylpropionic acid, beta-phenyl-l-alanine, h-phe-oh
PubChem CID 6140
ChEBI CHEBI:17295
IUPAC Name 2-amino-3-phenylpropanoic acid
SMILES NC(CC1=CC=CC=C1)C(O)=O

Specifications

Packaging Glass Bottle
Color White
Quantity 100 g
Physical Form Solid
Percent Purity ≥98.5% (Titration (dry basis))
Grade Biochemical
Chemical Name or Material L-Phenylalanine