missing translation for 'onlineSavingsMsg'
Learn More

L-Phenylalanine Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015027
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015027 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015027 Supplier Merck Emd Millipore Supplier No. PHR11001G

L-Phenylalanine is an essential aromatic amino acid. It has a role in metabolism as it forms a starting point for many pathways. This neutral amino acid is a precursor for tyrosine, epinephrine, dopamine and norepinephrine.

Specifications

CAS 63-91-2
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C6H5 CH2 CH(NH2)CO2 H
MDL Number MFCD00064227
Melting Point 270°C to 275°C
Synonym (S)-2-Amino-3-phenylpropionic acid
Type Certified Reference Material
Quantity 1 g
Formula Weight 165.19
Grade Certified Reference Material
Show More Show Less