missing translation for 'onlineSavingsMsg'
Learn More

L-Phenylalanine, 98.5%, Spectrum™ Chemical

Catalog No. 18611380
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611380 25 g Poly Pail
18611379 100 g Poly Pail
18611378 1 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18611380 Supplier Spectrum Chemical Mfg Cor Supplier No. PH14525GM
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 63-91-2

Specifications

Arsenic (As) 1.5 ppm
CAS Max % 1
Heavy Metals 20 ppm
Lead (Pb) 10 ppm
Loss on Drying 0.003
Packaging Poly Pail
Specific Rotation -33.2° to -35.2°
Quantity 25 g
Percent Purity 98.5%
Grade Reagent
Chemical Name or Material L-Phenylalanine
Show More Show Less

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.