missing translation for 'onlineSavingsMsg'
Learn More

L-Phenylalanine for Nitrogen Determination According to Kjeldahl Method, ≥99% (NT), MilliporeSigma™ Supelco™

Catalog No. 111015025
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015025 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015025 Supplier Merck Emd Millipore Supplier No. 9133125G

Specifications

CAS 63-91-2
Physical Form Neat
Linear Formula C6H5 CH2 CH(NH2)CO2 H
MDL Number MFCD00064227
Melting Point 270°C to 275°C
Synonym (S)-2-Amino-3-phenylpropionic acid
Solubility Soluble in water
Type Analytical Standard
Quantity 25 g
Formula Weight 165.19
Percent Purity ≥99% (NT)
Grade Analytical Standard
Show More Show Less