accessibility menu, dialog, popup

UserName

OHHS

CAS RN 60-24-2

IUPAC Name: 2-sulfanylethan-1-ol
Synonyms: β-mercaptoethanol monothioethyleneglycol thiomonoglycol thioglycol monothioglycol 2-mercaptoethanol (substance) mercaptoetanol
Molecular Weight (g/mol): 78.13
Molecular Formula: C2H6OS
InChi Key: DGVVWUTYPXICAM-UHFFFAOYSA-N
SMILES: OCCS
Quantity 
  • (3)
  • (2)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (9)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (4)
  • (1)
Physical Form 
  • (21)
Boiling Point 
  • (5)
  • (1)
  • (2)
  • (4)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (1)
  • (1)
  • (10)
  • (2)
  • (4)
  • (1)
  • (9)
  • (3)
  • (2)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (9)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (4)
  • (1)
  • (21)
  • (5)
  • (1)
  • (2)
  • (4)
  • (4)
  • (4)
  • (2)
  • (1)
  • (2)
  • (4)
  • (1)
  • (1)