missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptoethanol 98.0+%, TCI America™

Catalog No. M0058500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0058500G 500 g
M005825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0058500G Supplier TCI America Supplier No. M0058500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 60-24-2
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00004890
InChI Key DGVVWUTYPXICAM-UHFFFAOYSA-N
Synonym 2-mercaptoethanol, mercaptoethanol, thioglycol, beta-mercaptoethanol, ethanol, 2-mercapto, 2-thioethanol, 2-hydroxy-1-ethanethiol, thioethylene glycol, 2-hydroxyethanethiol, 2-hydroxyethyl mercaptan
PubChem CID 1567
ChEBI CHEBI:41218
IUPAC Name 2-sulfanylethan-1-ol
SMILES OCCS

Specifications

Melting Point -100°C
Color Colorless
Boiling Point 158°C
Quantity 500 g
UN Number 2966
Formula Weight 78.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Mercaptoethanol
EINECSNumber (2)-0458
RTECSNumber KL5600000
TSCA Yes