missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptoethanol Cell Culture MP Biomedicals

Catalog No. ICN19470591
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19470591 1 L
ICN19470580 100 mL
ICN19470583 250 mL
ICN19470590 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN19470591 Supplier MP Biomedicals Inc Supplier No. 0219470591

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 60-24-2
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00004890
InChI Key DGVVWUTYPXICAM-UHFFFAOYSA-N
Synonym 2-mercaptoethanol, mercaptoethanol, thioglycol, beta-mercaptoethanol, ethanol, 2-mercapto, 2-thioethanol, 2-hydroxy-1-ethanethiol, thioethylene glycol, 2-hydroxyethanethiol, 2-hydroxyethyl mercaptan
PubChem CID 1567
ChEBI CHEBI:41218
IUPAC Name 2-sulfanylethan-1-ol
SMILES OCCS

Specifications

Density 1.12g/mL
Assay Percent Range >98%
Quantity 1 L
Formula Weight 78.13
Grade Cell Culture Reagent
Physical Form Liquid
Chemical Name or Material 2-Mercaptoethanol, Molecular Biology and Proteomics
Recommended Storage Store at room temperature.