missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptoethanol, ≥98%, MP Biomedicals™

Catalog No. ICN19024291
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19024291 1 L
ICN19024280 100 mL
ICN19024283 250 mL
ICN19024290 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN19024291 Supplier MP Biomedicals Inc Supplier No. 0219024291

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 60-24-2
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00004890
InChI Key DGVVWUTYPXICAM-UHFFFAOYSA-N
Synonym 2-mercaptoethanol, mercaptoethanol, thioglycol, beta-mercaptoethanol, ethanol, 2-mercapto, 2-thioethanol, 2-hydroxy-1-ethanethiol, thioethylene glycol, 2-hydroxyethanethiol, 2-hydroxyethyl mercaptan
PubChem CID 1567
ChEBI CHEBI:41218
SMILES OCCS

Specifications

Color Colorless to Yellow
Density 1.12g/cm3 at 20°C
Assay Percent Range ≥98%
Quantity 1 L
IUPAC Name 2-sulfanylethan-1-ol
Formula Weight 78.13
Physical Form Liquid
Chemical Name or Material 2-Mercaptoethanol