missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ 2-Mercaptoethanol

Catalog No. PI35602
Change view
Click to view available options
Quantity:
10 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI35602 10 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI35602 Supplier Thermo Scientific™ Supplier No. 35602BID
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pure, liquid (equals 14M), beta-mercaptoethanol (bME, 2BME, 2-ME, b-mer), CAS 60-24-2; thiol reducing agent for cleaving protein disulfide bonds (cystines cysteines).

Thermo Scientific™ Pierce 2-Mercaptoethanol (2-ME), also called beta-mercaptoethanol (b-ME), is a mild reducing agent for cleaving protein disulfide bonds.

Highlights:

  • A mild reducing agent for cleaving disulfide bonds to thiols
  • Also known as β-Mercaptoethanol or BME
  • Often included in enzyme solutions to protect against catalytic site inactivation due to cysteine sulfhydryl oxidation/disulfide formation; added at final concentrations of 5 and 20mM, with or without EDTA as an additional protectant

Specifications

Concentration 14.3 M
CAS 60-24-2
Content And Storage Store between 2 to 25°C
Product Line Pierce
Form Liquid
Quantity 10 x 1 mL
Product Type Mercaptoethanol

For Research Use Only. Not for use in diagnostic procedures.