CAS RN 60-00-4
Ethylenediaminetetraacetic Acid (0.5M Solution/pH 8.0), Fisher BioReagents
CAS: 60-00-4 Molecular Formula: C10H16N2O8 Molecular Weight (g/mol): 292.24 MDL Number: MFCD00003541 InChI Key: KCXVZYZYPLLWCC-UHFFFAOYSA-N Synonym: edta,edetic acid,ethylenediaminetetraacetic acid,edathamil,versene,endrate,havidote,titriplex,edta acid,sequestrol PubChem CID: 6049 ChEBI: CHEBI:42191 IUPAC Name: 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid SMILES: OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
EDTA Titrant, 0.01M solution, Macron Fine Chemicals™
CAS: 60-00-4 Molecular Formula: C10H16N2O8 Molecular Weight (g/mol): 292.24 MDL Number: MFCD00003541 InChI Key: KCXVZYZYPLLWCC-UHFFFAOYSA-N Synonym: edta,edetic acid,ethylenediaminetetraacetic acid,edathamil,versene,endrate,havidote,titriplex,edta acid,sequestrol PubChem CID: 6049 ChEBI: CHEBI:42191 IUPAC Name: 2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid SMILES: OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
Ethylenediamine Tetraacetic Acid (Certified ACS), Fisher Chemical™
CAS: 60-00-4 Molecular Formula: C10H16N2O8 Molecular Weight (g/mol): 292.24 MDL Number: MFCD00003541 InChI Key: KCXVZYZYPLLWCC-UHFFFAOYSA-N Synonym: edta,edetic acid,ethylenediaminetetraacetic acid,edathamil,versene,endrate,havidote,titriplex,edta acid,sequestrol PubChem CID: 6049 ChEBI: CHEBI:42191 IUPAC Name: 2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid SMILES: OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
EDTA Titrant, 0.05M solution, Macron Fine Chemicals™
CAS: 60-00-4 Molecular Formula: C10H16N2O8 Molecular Weight (g/mol): 292.24 MDL Number: MFCD00003541 InChI Key: KCXVZYZYPLLWCC-UHFFFAOYSA-N Synonym: edta,edetic acid,ethylenediaminetetraacetic acid,edathamil,versene,endrate,havidote,titriplex,edta acid,sequestrol PubChem CID: 6049 ChEBI: CHEBI:42191 IUPAC Name: 2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid SMILES: OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
Thermo Scientific Chemicals Ethylenediaminetetraacetic acid, 0.5M aq. soln, pH 8.0
CAS: 60-00-4 Molecular Formula: C10H16N2O8 Molecular Weight (g/mol): 292.24 MDL Number: MFCD00003541 InChI Key: KCXVZYZYPLLWCC-UHFFFAOYSA-N Synonym: edta,edetic acid,ethylenediaminetetraacetic acid,edathamil,versene,endrate,havidote,titriplex,edta acid,sequestrol PubChem CID: 6049 ChEBI: CHEBI:42191 IUPAC Name: 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid SMILES: OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
Thermo Scientific Chemicals Ethylenediaminetetraacetic Acid, 0.5M aq. soln., pH 8.0, autoclaved
CAS: 60-00-4 Molecular Formula: C10H16N2O8 Molecular Weight (g/mol): 292.24 MDL Number: MFCD00003541 InChI Key: KCXVZYZYPLLWCC-UHFFFAOYSA-N Synonym: edta,edetic acid,ethylenediaminetetraacetic acid,edathamil,versene,endrate,havidote,titriplex,edta acid,sequestrol PubChem CID: 6049 ChEBI: CHEBI:42191 IUPAC Name: 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid SMILES: OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
MilliporeSigma™ EDTA, OmniPur™, Calbiochem™,
CAS: 60-00-4 Molecular Formula: C10H16N2O8 Molecular Weight (g/mol): 292.24 MDL Number: MFCD00003541 InChI Key: KCXVZYZYPLLWCC-UHFFFAOYSA-N Synonym: edta,edetic acid,ethylenediaminetetraacetic acid,edathamil,versene,endrate,havidote,titriplex,edta acid,sequestrol PubChem CID: 6049 ChEBI: CHEBI:42191 IUPAC Name: 2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid SMILES: OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
Thermo Scientific Chemicals Ethylenediaminetetraacetic acid, Electrophoresis Grade, 99.4+%
CAS: 60-00-4 Molecular Formula: C10H16N2O8 Molecular Weight (g/mol): 292.24 MDL Number: MFCD00003541 InChI Key: KCXVZYZYPLLWCC-UHFFFAOYSA-N Synonym: edta,edetic acid,ethylenediaminetetraacetic acid,edathamil,versene,endrate,havidote,titriplex,edta acid,sequestrol PubChem CID: 6049 ChEBI: CHEBI:42191 SMILES: OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
Thermo Scientific Chemicals Ethylenediaminetetraacetic acid, 99%, pure
CAS: 60-00-4 Molecular Formula: C10H16N2O8 Molecular Weight (g/mol): 292.24 MDL Number: MFCD00003541 InChI Key: KCXVZYZYPLLWCC-UHFFFAOYSA-N Synonym: edta,edetic acid,ethylenediaminetetraacetic acid,edathamil,versene,endrate,havidote,titriplex,edta acid,sequestrol PubChem CID: 6049 ChEBI: CHEBI:42191 IUPAC Name: 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid SMILES: OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
Thermo Scientific Chemicals Ethylenediaminetetraacetic acid, Cell Culture Reagent
CAS: 60-00-4 Molecular Formula: C10H16N2O8 Molecular Weight (g/mol): 292.24 MDL Number: MFCD00003541 InChI Key: KCXVZYZYPLLWCC-UHFFFAOYSA-N Synonym: edta,edetic acid,ethylenediaminetetraacetic acid,edathamil,versene,endrate,havidote,titriplex,edta acid,sequestrol PubChem CID: 6049 ChEBI: CHEBI:42191 SMILES: OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
MilliporeSigma™ Calbiochem™ EDTA, 500mM Solution, pH 8.0, ULTROL™ Grade
Application in Life Science research
Thermo Scientific Chemicals Ethylenediaminetetraacetic acid, 99.5%, ACS reagent
CAS: 60-00-4 Molecular Formula: C10H16N2O8 Molecular Weight (g/mol): 292.24 MDL Number: MFCD00003541 InChI Key: KCXVZYZYPLLWCC-UHFFFAOYSA-N Synonym: edta,edetic acid,ethylenediaminetetraacetic acid,edathamil,versene,endrate,havidote,titriplex,edta acid,sequestrol PubChem CID: 6049 ChEBI: CHEBI:42191 IUPAC Name: 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid SMILES: OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
Ethylenediaminetetraacetic acid, ACS, 99.4+%
CAS: 60-00-4 Molecular Formula: C10H16N2O8 Molecular Weight (g/mol): 292.24 MDL Number: MFCD00003541 InChI Key: KCXVZYZYPLLWCC-UHFFFAOYSA-N Synonym: edta,edetic acid,ethylenediaminetetraacetic acid,edathamil,versene,endrate,havidote,titriplex,edta acid,sequestrol PubChem CID: 6049 ChEBI: CHEBI:42191 IUPAC Name: 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid SMILES: OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
Ethylenediaminetetraacetic acid, 99%
CAS: 60-00-4 Molecular Formula: C10H16N2O8 Molecular Weight (g/mol): 292.24 MDL Number: MFCD00003541 InChI Key: KCXVZYZYPLLWCC-UHFFFAOYSA-N Synonym: edta,edetic acid,ethylenediaminetetraacetic acid,edathamil,versene,endrate,havidote,titriplex,edta acid,sequestrol PubChem CID: 6049 ChEBI: CHEBI:42191 IUPAC Name: 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid SMILES: OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
- 1
- 2