missing translation for 'onlineSavingsMsg'
Learn More

Ethylenedinitrilo)Tetraacetic Acid ACS AR Powder, Macron Fine Chemicals™

Catalog No. MK2580500
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MK2580500 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK2580500 Supplier Avantor Supplier No. 258012

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 60-00-4
Packaging POLYSTORMOR
Molecular Formula C10H16N2O8
MDL Number MFCD00003541
Quantity 500 g
Synonym edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol
InChI Key KCXVZYZYPLLWCC-UHFFFAOYSA-N
SMILES OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
IUPAC Name 2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid
Molecular Weight (g/mol) 292.24
PubChem CID 6049
ChEBI CHEBI:42191
Formula Weight 292.25
Grade ACS
Physical Form Powder
Chemical Name or Material (Ethylenedinitrilo)Tetraacetic Acid
Show More Show Less