missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ EDTA, 500mM Solution, pH 8.0, ULTROL™ Grade

Catalog No. 324504500ML
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
324504500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 324504500ML Supplier MilliporeSigma™ Supplier No. 324504500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Application in Life Science research

  • Colorless to pale yellow solution
  • Irritating to eyes and skin

Specifications

CAS 60-00-4
Content And Storage 15°C to 30°C
Form Solution
Product Type EDTA
Quantity 500 mL