missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ethylenediaminetetraacetic acid, 99.5%, ACS reagent

Catalog No. AC409930050
Change view
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC409930050 5 kg Plastic bottle
AC409930010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC409930050 Supplier Thermo Scientific Chemicals Supplier No. 409930050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 60-00-4 | C10H16N2O8 | 292.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 60-00-4
Molecular Formula C10H16N2O8
Molecular Weight (g/mol) 292.24
MDL Number MFCD00003541
InChI Key KCXVZYZYPLLWCC-UHFFFAOYSA-N
Synonym edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol
PubChem CID 6049
ChEBI CHEBI:42191
IUPAC Name 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid
SMILES OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

Specifications

Decomposition Information >150°C
Melting Point 220.0°C
Color White
Flash Point >100°C
Ignition Residue 0.2% max.
Infrared Spectrum Authentic
Assay Percent Range 99.4 to 100.6% (Complexometry)
Packaging Plastic bottle
Linear Formula (HO2CCH2)2NCH2CH2N(CH2CO2H)2
Quantity 5 kg
Fieser 01,373
Merck Index 15, 3565
Solubility Information Solubility in water: 0.5g/L (20°C) - 2.2g/L (80°C). Other solubilities: insoluble in common organic solvents, insoluble in dilute ammonium hydroxide: 0.005% max
Vapor Pressure 0.013hPa at 20°C
Formula Weight 292.23
Percent Purity 99.5%
Grade ACS Reagent
Insoluble Matter 0.005% max. (in dilute NH4OH)
Physical Form Fine Crystalline Powder
Chemical Name or Material Ethylenediaminetetraacetic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cau
EINECSNumber 200-449-4
RTECSNumber AH4025000
TSCA TSCA

RUO – Research Use Only