CAS RN 599-79-1
Sulfasalazine, 97%
CAS: 599-79-1 Molecular Formula: C18H14N4O5S Molecular Weight (g/mol): 398.39 InChI Key: OQANPHBRHBJGNZ-BKUYFWCQSA-N IUPAC Name: (3Z)-6-oxo-3-(2-{4-[(pyridin-2-yl)sulfamoyl]phenyl}hydrazin-1-ylidene)cyclohexa-1,4-diene-1-carboxylic acid SMILES: OC(=O)C1=C\C(C=CC1=O)=N/NC1=CC=C(C=C1)S(=O)(=O)NC1=CC=CC=N1
Sulfasalazine, 98.0-101.5%, MilliporeSigma™ Supelco™
Chemical structure: sulfonamide
Sulfasalazine, 97%, Thermo Scientific™
CAS: 599-79-1 Molecular Formula: C18H14N4O5S Molecular Weight (g/mol): 398.39 InChI Key: OQANPHBRHBJGNZ-BKUYFWCQSA-N IUPAC Name: (3Z)-6-oxo-3-(2-{4-[(pyridin-2-yl)sulfamoyl]phenyl}hydrazin-1-ylidene)cyclohexa-1,4-diene-1-carboxylic acid SMILES: OC(=O)C1=C\C(C=CC1=O)=N/NC1=CC=C(C=C1)S(=O)(=O)NC1=CC=CC=N1
Sulfasalazine 95.0+%, TCI America™
CAS: 599-79-1 Molecular Formula: C18H14N4O5S Molecular Weight (g/mol): 398.393 MDL Number: MFCD00057363 InChI Key: OQANPHBRHBJGNZ-UHFFFAOYSA-N Synonym: sulfasalazine,salazosulfapyridine,azulfidine,salicylazosulfapyridine,salazopyridin,salazopyrin,asulfidine,sulfazalazine,sulphasalazine,accucol PubChem CID: 5384001 IUPAC Name: 6-oxo-3-[[4-(pyridin-2-ylsulfamoyl)phenyl]hydrazinylidene]cyclohexa-1,4-diene-1-carboxylic acid SMILES: C1=CC=NC(=C1)NS(=O)(=O)C2=CC=C(C=C2)NN=C3C=CC(=O)C(=C3)C(=O)O
R&D Systems™ Sulfasalazine
CAS: 599-79-1 Molecular Formula: C18H14N4O5S