missing translation for 'onlineSavingsMsg'
Learn More

Sulfasalazine USP , Medisca™
GREENER_CHOICE

Catalog No. 18048124
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18048124 500 g
18048028 25 g
18048029 1 kg
18048084 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18048124 Supplier Medisca Pharmaceutique Inc Supplier No. 017608
Get 1 Extra Chemical When You Buy 4
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sulfasalazine USP is a bright yellow or brownish yellow, odorless, fine powder.

Chemical Identifiers

CAS 599-79-1

Specifications

Type α-Carboxybenzyl Penicillin Disodium Salt
Physical Form Powder
Color Brown to Yellow
Quantity 500 g
Percent Purity 97.0 to 101.5%
Grade USP