missing translation for 'onlineSavingsMsg'
Learn More

Sulfasalazine 95.0+%, TCI America™

Catalog No. S058025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S058025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S058025G Supplier TCI America Supplier No. S058025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 599-79-1
Molecular Formula C18H14N4O5S
Molecular Weight (g/mol) 398.393
MDL Number MFCD00057363
InChI Key OQANPHBRHBJGNZ-UHFFFAOYSA-N
Synonym sulfasalazine, salazosulfapyridine, azulfidine, salicylazosulfapyridine, salazopyridin, salazopyrin, asulfidine, sulfazalazine, sulphasalazine, accucol
PubChem CID 5384001
IUPAC Name 6-oxo-3-[[4-(pyridin-2-ylsulfamoyl)phenyl]hydrazinylidene]cyclohexa-1,4-diene-1-carboxylic acid
SMILES C1=CC=NC(=C1)NS(=O)(=O)C2=CC=C(C=C2)NN=C3C=CC(=O)C(=C3)C(=O)O

Specifications

Melting Point 246°C
Quantity 25 g
Formula Weight 398.39
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sulfasalazine
EINECSNumber (9)-0483
RTECSNumber VO6250000
TSCA Yes