missing translation for 'onlineSavingsMsg'
Learn More

Sulfasalazine, 97%, Thermo Scientific™

Catalog No. p-7214389
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461240250 25 g
AC461241000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC461240250 Supplier Thermo Scientific Chemicals Supplier No. 461240250

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 599-79-1
Molecular Formula C18H14N4O5S
Molecular Weight (g/mol) 398.39
InChI Key OQANPHBRHBJGNZ-BKUYFWCQSA-N
IUPAC Name (3Z)-6-oxo-3-(2-{4-[(pyridin-2-yl)sulfamoyl]phenyl}hydrazin-1-ylidene)cyclohexa-1,4-diene-1-carboxylic acid
SMILES OC(=O)C1=C\C(C=CC1=O)=N/NC1=CC=C(C=C1)S(=O)(=O)NC1=CC=CC=N1

Specifications

Assay Percent Range 96.0 % min. (HPLC)
Quantity 25 g
Solubility Information Solubility aq. soln.: insoluble. Other solubilities: slightly soluble in alcohols, insoluble in ether, benzene, chloroform
Formula Weight 398.39
Chemical Name or Material Sulfasalazine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement May cause an allergic skin reaction.

May cause allergy or asthma symptoms or breathing difficulties if inhaled.

Suspected of causing cancer.

Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water.

In case of inadequate ventilation wear respiratory protection.

IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.

If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.

Wear protective gloves/protective clothing/eye protection/face protection.

IF exposed or concerned: Get medical advice/attention.

EINECSNumber 209-974-3

RUO – Research Use Only