CAS RN 5965-83-3
5-Sulfosalicylic Acid Dihydrate (Crystalline/Certified ACS), Fisher Chemical™
CAS: 5965-83-3 Molecular Formula: C9H14O6S Molecular Weight (g/mol): 250.27 MDL Number: MFCD00007508,MFCD00149540 InChI Key: NFYHZVWMQHQKRU-UHFFFAOYSA-N Synonym: 2-hydroxy-5-sulfobenzoic acid dihydrate,5-sulfosalicylic acid dihydrate,sulfosalicylic acid dihydrate,benzoic acid, 2-hydroxy-5-sulfo-, dihydrate,unii-09ngq462s6,2-hydroxy-5-sulfobenzoic acid, hydrate, hydrate,5-sulfosalicylsyre,5-sulfosalicylsaeure,salicylic acid, 5-sulfo-, dihydrate,acmc-1aymd PubChem CID: 2723734 IUPAC Name: 2-hydroxy-5-sulfobenzoic acid; bis(methane) SMILES: C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O
5-Sulfosalicylic acid dihydrate, 98%
CAS: 5965-83-3 Molecular Formula: C9H14O6S Molecular Weight (g/mol): 250.27 MDL Number: MFCD00007508,MFCD00149540 InChI Key: NFYHZVWMQHQKRU-UHFFFAOYSA-N Synonym: 2-hydroxy-5-sulfobenzoic acid dihydrate,5-sulfosalicylic acid dihydrate,sulfosalicylic acid dihydrate,benzoic acid, 2-hydroxy-5-sulfo-, dihydrate,unii-09ngq462s6,2-hydroxy-5-sulfobenzoic acid, hydrate, hydrate,5-sulfosalicylsyre,5-sulfosalicylsaeure,salicylic acid, 5-sulfo-, dihydrate,acmc-1aymd PubChem CID: 2723734 SMILES: C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O
Thermo Scientific Chemicals 5-Sulfosalicylic acid dihydrate, 99+%
CAS: 5965-83-3 | C9H14O6S | 250.27 g/mol
Thermo Scientific™ 5-Sulfosalicylic acid dihydrate, 99+%
5-Sulfosalicylic acid dihydrate, ACS, 99+%
CAS: 5965-83-3 Molecular Formula: C9H14O6S Molecular Weight (g/mol): 250.27 MDL Number: MFCD00007508,MFCD00149540 InChI Key: NFYHZVWMQHQKRU-UHFFFAOYSA-N Synonym: 2-hydroxy-5-sulfobenzoic acid dihydrate,5-sulfosalicylic acid dihydrate,sulfosalicylic acid dihydrate,benzoic acid, 2-hydroxy-5-sulfo-, dihydrate,unii-09ngq462s6,2-hydroxy-5-sulfobenzoic acid, hydrate, hydrate,5-sulfosalicylsyre,5-sulfosalicylsaeure,salicylic acid, 5-sulfo-, dihydrate,acmc-1aymd PubChem CID: 2723734 IUPAC Name: 2-hydroxy-5-sulfobenzoic acid;dihydrate SMILES: C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O
5-Sulfosalicylic acid dihydrate, ACS reagent
CAS: 5965-83-3 Molecular Formula: C9H14O6S Molecular Weight (g/mol): 250.27 MDL Number: MFCD00007508,MFCD00149540 InChI Key: NFYHZVWMQHQKRU-UHFFFAOYSA-N Synonym: 2-hydroxy-5-sulfobenzoic acid dihydrate,5-sulfosalicylic acid dihydrate,sulfosalicylic acid dihydrate,benzoic acid, 2-hydroxy-5-sulfo-, dihydrate,unii-09ngq462s6,2-hydroxy-5-sulfobenzoic acid, hydrate, hydrate,5-sulfosalicylsyre,5-sulfosalicylsaeure,salicylic acid, 5-sulfo-, dihydrate,acmc-1aymd PubChem CID: 2723734 IUPAC Name: 2-hydroxy-5-sulfobenzoic acid;dihydrate SMILES: C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O
Sulfosalicylic Acid, Dihydrate, ACS Grade, 99.0 to 101.0%, LabChem™
CAS: 5965-83-3 Molecular Formula: C9H14O6S Molecular Weight (g/mol): 250.27 MDL Number: MFCD00007508,MFCD00149540 InChI Key: NFYHZVWMQHQKRU-UHFFFAOYSA-N Synonym: 2-hydroxy-5-sulfobenzoic acid dihydrate,5-sulfosalicylic acid dihydrate,sulfosalicylic acid dihydrate,benzoic acid, 2-hydroxy-5-sulfo-, dihydrate,unii-09ngq462s6,2-hydroxy-5-sulfobenzoic acid, hydrate, hydrate,5-sulfosalicylsyre,5-sulfosalicylsaeure,salicylic acid, 5-sulfo-, dihydrate,acmc-1aymd PubChem CID: 2723734 IUPAC Name: 2-hydroxy-5-sulfobenzoic acid; bis(methane) SMILES: C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O
Sulfosalicylic Acid, Certified, 3.0% (w/v) ±0.1%, LabChem™
CAS: 5965-83-3 Molecular Formula: C9H14O6S Molecular Weight (g/mol): 250.27 MDL Number: MFCD00007508,MFCD00149540 InChI Key: NFYHZVWMQHQKRU-UHFFFAOYSA-N Synonym: 2-hydroxy-5-sulfobenzoic acid dihydrate,5-sulfosalicylic acid dihydrate,sulfosalicylic acid dihydrate,benzoic acid, 2-hydroxy-5-sulfo-, dihydrate,unii-09ngq462s6,2-hydroxy-5-sulfobenzoic acid, hydrate, hydrate,5-sulfosalicylsyre,5-sulfosalicylsaeure,salicylic acid, 5-sulfo-, dihydrate,acmc-1aymd PubChem CID: 2723734 IUPAC Name: 2-hydroxy-5-sulfobenzoic acid; bis(methane) SMILES: C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O
Sulfosalicylic Acid, Certified, 20.0% (w/v) ±0.5%, LabChem™
CAS: 5965-83-3 Molecular Formula: C9H14O6S Molecular Weight (g/mol): 250.27 MDL Number: MFCD00007508,MFCD00149540 InChI Key: NFYHZVWMQHQKRU-UHFFFAOYSA-N Synonym: 2-hydroxy-5-sulfobenzoic acid dihydrate,5-sulfosalicylic acid dihydrate,sulfosalicylic acid dihydrate,benzoic acid, 2-hydroxy-5-sulfo-, dihydrate,unii-09ngq462s6,2-hydroxy-5-sulfobenzoic acid, hydrate, hydrate,5-sulfosalicylsyre,5-sulfosalicylsaeure,salicylic acid, 5-sulfo-, dihydrate,acmc-1aymd PubChem CID: 2723734 IUPAC Name: 2-hydroxy-5-sulfobenzoic acid; bis(methane) SMILES: C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O
Sulfosalicylic Acid, 10% (w/v), Ricca Chemical
CAS: 5965-83-3 Molecular Formula: C9H14O6S Molecular Weight (g/mol): 250.27 MDL Number: MFCD00007508,MFCD00149540 InChI Key: NFYHZVWMQHQKRU-UHFFFAOYSA-N Synonym: 2-hydroxy-5-sulfobenzoic acid dihydrate,5-sulfosalicylic acid dihydrate,sulfosalicylic acid dihydrate,benzoic acid, 2-hydroxy-5-sulfo-, dihydrate,unii-09ngq462s6,2-hydroxy-5-sulfobenzoic acid, hydrate, hydrate,5-sulfosalicylsyre,5-sulfosalicylsaeure,salicylic acid, 5-sulfo-, dihydrate,acmc-1aymd PubChem CID: 2723734 IUPAC Name: 2-hydroxy-5-sulfobenzoic acid; bis(methane) SMILES: C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O
Sulfosalicylic Acid, Certified, 5.0% (w/v) ±0.1%, LabChem™
CAS: 5965-83-3 Molecular Formula: C9H14O6S Molecular Weight (g/mol): 250.27 MDL Number: MFCD00007508,MFCD00149540 InChI Key: NFYHZVWMQHQKRU-UHFFFAOYSA-N Synonym: 2-hydroxy-5-sulfobenzoic acid dihydrate,5-sulfosalicylic acid dihydrate,sulfosalicylic acid dihydrate,benzoic acid, 2-hydroxy-5-sulfo-, dihydrate,unii-09ngq462s6,2-hydroxy-5-sulfobenzoic acid, hydrate, hydrate,5-sulfosalicylsyre,5-sulfosalicylsaeure,salicylic acid, 5-sulfo-, dihydrate,acmc-1aymd PubChem CID: 2723734 IUPAC Name: 2-hydroxy-5-sulfobenzoic acid; bis(methane) SMILES: C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O
Sulfosalicylic Acid, 5% (w/v), Ricca Chemical
CAS: 5965-83-3 Molecular Formula: C9H14O6S Molecular Weight (g/mol): 250.27 MDL Number: MFCD00007508,MFCD00149540 InChI Key: NFYHZVWMQHQKRU-UHFFFAOYSA-N Synonym: 2-hydroxy-5-sulfobenzoic acid dihydrate,5-sulfosalicylic acid dihydrate,sulfosalicylic acid dihydrate,benzoic acid, 2-hydroxy-5-sulfo-, dihydrate,unii-09ngq462s6,2-hydroxy-5-sulfobenzoic acid, hydrate, hydrate,5-sulfosalicylsyre,5-sulfosalicylsaeure,salicylic acid, 5-sulfo-, dihydrate,acmc-1aymd PubChem CID: 2723734 IUPAC Name: 2-hydroxy-5-sulfobenzoic acid; bis(methane) SMILES: C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O
- 1
- 2