missing translation for 'onlineSavingsMsg'
Learn More

Sulfosalicylic Acid, Certified, 20.0% (w/v) ±0.5%, LabChem™

Catalog No. LC255451
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Amber Poly
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC255451 500 mL Amber Poly
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC255451 Supplier LabChem Supplier No. LC255451

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5965-83-3 , 7732-18-5
Molecular Formula C9H14O6S
Molecular Weight (g/mol) 250.27
MDL Number MFCD00007508,MFCD00149540
InChI Key NFYHZVWMQHQKRU-UHFFFAOYSA-N
Synonym 2-hydroxy-5-sulfobenzoic acid dihydrate, 5-sulfosalicylic acid dihydrate, sulfosalicylic acid dihydrate, benzoic acid, 2-hydroxy-5-sulfo-, dihydrate, unii-09ngq462s6, 2-hydroxy-5-sulfobenzoic acid, hydrate, hydrate, 5-sulfosalicylsyre, 5-sulfosalicylsaeure, salicylic acid, 5-sulfo-, dihydrate, acmc-1aymd
PubChem CID 2723734
IUPAC Name 2-hydroxy-5-sulfobenzoic acid; bis(methane)
SMILES C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O

Specifications

CAS Max % 80, 20
Color Pink
Packaging Amber Poly
Linear Formula C7H6O6S·2H2O
Quantity 500 mL
Solubility Information Soluble in water
Formula Weight 254.21
Percent Purity 20.0% ±0.5% (w/v)
Grade Certified
Physical Form Liquid
Chemical Name or Material Sulfosalicylic Acid
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature