missing translation for 'onlineSavingsMsg'
Learn More

Sulfosalicylic Acid, 10% (w/v), Ricca Chemical

Catalog No. 81251
Change view
Click to view available options
Package: Amber Glass; Size: 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
81251 Package: Amber Glass; Size: 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 81251 Supplier Ricca Chemical Company Supplier No. 81251

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 5965-83-3, 7732-18-5
CAS Min % 89.0, 8.91
CAS Max % 93.0, 9.28
Molecular Formula C9H14O6S
MDL Number MFCD00007508,MFCD00149540
Synonym 2-hydroxy-5-sulfobenzoic acid dihydrate, 5-sulfosalicylic acid dihydrate, sulfosalicylic acid dihydrate, benzoic acid, 2-hydroxy-5-sulfo-, dihydrate, unii-09ngq462s6, 2-hydroxy-5-sulfobenzoic acid, hydrate, hydrate, 5-sulfosalicylsyre, 5-sulfosalicylsaeure, salicylic acid, 5-sulfo-, dihydrate, acmc-1aymd
InChI Key NFYHZVWMQHQKRU-UHFFFAOYSA-N
SMILES C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O
IUPAC Name 2-hydroxy-5-sulfobenzoic acid; bis(methane)
Molecular Weight (g/mol) 250.27
PubChem CID 2723734
Concentration or Composition (by Analyte or Components) 10%(w/v)
Packaging Amber Glass Bottle
Solvent or Matrix Aqueous solution
Quantity 4 L
Show More Show Less