missing translation for 'onlineSavingsMsg'
Learn More

Sulfosalicylic Acid, 10% (w/v), Ricca Chemical

Catalog No. 812516
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
812516 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 812516 Supplier Ricca Chemical Company Supplier No. 812516

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5965-83-3 , 7732-18-5
Molecular Formula C9H14O6S
Molecular Weight (g/mol) 250.27
MDL Number MFCD00007508,MFCD00149540
InChI Key NFYHZVWMQHQKRU-UHFFFAOYSA-N
Synonym 2-hydroxy-5-sulfobenzoic acid dihydrate, 5-sulfosalicylic acid dihydrate, sulfosalicylic acid dihydrate, benzoic acid, 2-hydroxy-5-sulfo-, dihydrate, unii-09ngq462s6, 2-hydroxy-5-sulfobenzoic acid, hydrate, hydrate, 5-sulfosalicylsyre, 5-sulfosalicylsaeure, salicylic acid, 5-sulfo-, dihydrate, acmc-1aymd
PubChem CID 2723734
IUPAC Name 2-hydroxy-5-sulfobenzoic acid; bis(methane)
SMILES C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O

Specifications

CAS Min % 89.0, 8.91
CAS Max % 93.0, 9.28
Percent Purity 10%
Grade Laboratory
Concentration or Composition (by Analyte or Components) 10%(w/v)
Packaging Amber Glass Bottle
Solvent or Matrix Aqueous solution
Quantity 500 mL
Chemical Name or Material Sulfosalicylic Acid