missing translation for 'onlineSavingsMsg'
Learn More

Sulfosalicylic Acid, Certified, 3.0% (w/v) ±0.1%, LabChem™

Catalog No. LC255304
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Amber Poly
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC255304 4 L Poly Bottle
LC255301 500 mL Amber Poly
LC255302 1 L Amber Poly
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC255304 Supplier LabChem Supplier No. LC255304

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5965-83-3 , 7732-18-5
Molecular Formula C9H14O6S
Molecular Weight (g/mol) 250.27
MDL Number MFCD00007508,MFCD00149540
InChI Key NFYHZVWMQHQKRU-UHFFFAOYSA-N
Synonym 2-hydroxy-5-sulfobenzoic acid dihydrate, 5-sulfosalicylic acid dihydrate, sulfosalicylic acid dihydrate, benzoic acid, 2-hydroxy-5-sulfo-, dihydrate, unii-09ngq462s6, 2-hydroxy-5-sulfobenzoic acid, hydrate, hydrate, 5-sulfosalicylsyre, 5-sulfosalicylsaeure, salicylic acid, 5-sulfo-, dihydrate, acmc-1aymd
PubChem CID 2723734
IUPAC Name 2-hydroxy-5-sulfobenzoic acid; bis(methane)
SMILES C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O

Specifications

CAS Max % 97, 3
Color Colorless
Packaging Poly Bottle
Linear Formula C7H6O6S·2H2O
Quantity 4 L
Solubility Information Soluble in water
Formula Weight 254.21
Percent Purity 3.0% ±0.1% (w/v)
Grade Certified
Physical Form Liquid
Chemical Name or Material Sulfosalicylic Acid
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Recommended Storage Room Temperature