accessibility menu, dialog, popup

UserName

NH2OHOHO

CAS RN 56-45-1

IUPAC Name: 2-amino-3-hydroxypropanoic acid
Synonyms: serin DSN serine DL-form serine D-form
Molecular Weight (g/mol): 105.09
Molecular Formula: C3H7NO3
InChi Key: MTCFGRXMJLQNBG-UHFFFAOYNA-N
SMILES: NC(CO)C(O)=O
Molecular Weight (g/mol) 
  • (11)
  • (15)
Quantity 
  • (4)
  • (1)
  • (7)
  • (1)
  • (1)
  • (6)
  • (1)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
  • (8)
  • (3)
  • (7)
Grade 
  • (1)
  • (4)
  • (1)
  • (3)
  • (3)
  • (1)
  • (5)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (18)
  • (2)
  • (1)
  • (11)
  • (3)
  • (11)
  • (11)
  • (15)
  • (4)
  • (1)
  • (7)
  • (1)
  • (1)
  • (6)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (8)
  • (3)
  • (7)
  • (1)
  • (4)
  • (1)
  • (3)
  • (3)
  • (1)
  • (5)
  • (3)