missing translation for 'onlineSavingsMsg'
Learn More

Serine, USP, 98.5-101.5%, Spectrum™ Chemical

Catalog No. 18606185
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
12 kg
25 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606185 12 kg Poly Bottle
18606181 25 g Poly Bottle
18606182 100 g Poly Bottle
18606183 1 kg Poly Bottle
18606184 25 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18606185 Supplier Spectrum Chemical Mfg Cor Supplier No. S102312KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

S1023, 56-45-1, C3H7NO3

Spectrum™ Chemical Serine, USP - All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 56-45-1
Molecular Formula C3H7NO3
Molecular Weight (g/mol) 105.09
InChI Key MTCFGRXMJLQNBG-UHFFFAOYNA-N
IUPAC Name 2-amino-3-hydroxypropanoic acid
SMILES NC(CO)C(O)=O

Specifications

CAS Max % 1
Ignition Residue 0.001
Loss on Drying 0.002
Packaging Poly Bottle
Quantity 12 kg
Specific Rotation +14.0° to +15.6°
Percent Purity 98.5 to 101.5%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Serine