missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Serine, 99%

Catalog No. AC132665000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC132665000 500 g Glass Bottle
AC132660250 25 g Glass Bottle
AC132661000 100 g Glass Bottle
AC132660025 2.5 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC132665000 Supplier Thermo Scientific Chemicals Supplier No. 132665000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 56-45-1 | C3H7NO3 | 105.093 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56-45-1
Molecular Formula C3H7NO3
Molecular Weight (g/mol) 105.093
InChI Key MTCFGRXMJLQNBG-REOHCLBHSA-N
Synonym l-serine, serine, s-2-amino-3-hydroxypropanoic acid, s-serine, beta-hydroxyalanine, h-ser-oh, l-ser, l---serine, serine, l, 2s-2-amino-3-hydroxypropanoic acid
PubChem CID 5951
ChEBI CHEBI:17115
IUPAC Name (2S)-2-amino-3-hydroxypropanoic acid
SMILES C(C(C(=O)O)N)O

Specifications

Melting Point 222.0°C
Ammonium 200ppm max.
Arsenic (As) 1ppm max.
pH 5.0 to 6.5 (2.5% soln. at 25°C)
Color White
Chlorine (Cl) 200ppm max.
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Iron (Fe) 10ppm max.
Loss on Drying 0.2% (105°C, 3 hrs) max.
Assay Percent Range 99%
Other Amino Acids 0.5% (TLC) max.
Packaging Glass Bottle
Linear Formula HOCH2CH(NH2)CO2H
Specific Rotation 15.2
Sulfate 200ppm max.
Transmittance 95% (c=10, H2O, 430nm) 1cm cellmin.
Sulfated Ash 0.1% max.
Solubility Information Solubility in water: 250g/L (20°C). Other solubilities: insoluble in alcohol, ether and benzene, soluble in mineral acids, insoluble in organic solvents, insoluble in ethanol and diethylether
Sublimation Point 0150.0°C
Density 1.6000g/mL
Specific Rotation Condition 15.20 (20.00°C c=10,2N HCl)
Quantity 500 g
Formula Weight 105.09
Percent Purity 99%
Specific Gravity 1.6
Physical Form Crystalline Powder, Crystals or Crystalline Needles
Chemical Name or Material L-Serine
Show More Show Less

RUO – Research Use Only