missing translation for 'onlineSavingsMsg'
Learn More

L-Serine, FCC, 98.5-101.5%, Spectrum™ Chemical

Catalog No. 18603981
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603981 1 kg Poly Bottle
18603979 25 g Poly Bottle
18603980 100 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18603981 Supplier Spectrum Chemical Mfg Cor Supplier No. S10211KG
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

S1021, 56-45-1, C3H7NO3

Spectrum™ Chemical L-Serine, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 56-45-1
Molecular Formula C3H7NO3
Molecular Weight (g/mol) 105.09
InChI Key MTCFGRXMJLQNBG-UHFFFAOYNA-N
IUPAC Name 2-amino-3-hydroxypropanoic acid
SMILES NC(CO)C(O)=O

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 0.3%
Packaging Poly Bottle
Quantity 1 kg
Optical Rotation +13.6° to +160°
Percent Purity 98.5 to 101.5%
Grade FCC
Physical Form Crystalline Powder or Lumps