missing translation for 'onlineSavingsMsg'
Learn More

L-Serine Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014643
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014643 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014643 Supplier Merck Emd Millipore Supplier No. PHR11031G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 56-45-1
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula HOCH2 CH(NH2)CO2 H
MDL Number MFCD00064224
Melting Point 222°C
Synonym (S)-2-Amino-3-hydroxypropionic acid
Type Certified Reference Material
Quantity 1 g
Formula Weight 105.09
Grade Certified Reference Material
Show More Show Less