missing translation for 'onlineSavingsMsg'
Learn More

L-Serine 99.0+%, TCI America™

Catalog No. S00355G
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S00355G 5 g
S003525G 25 g
S0035250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S00355G Supplier TCI America Supplier No. S00355G

Chemical Identifiers

CAS 56-45-1
Molecular Formula C3H7NO3
Molecular Weight (g/mol) 105.093
MDL Number MFCD00064224
InChI Key MTCFGRXMJLQNBG-REOHCLBHSA-N
Synonym l-serine, serine, s-2-amino-3-hydroxypropanoic acid, s-serine, beta-hydroxyalanine, h-ser-oh, l-ser, l---serine, serine, l, 2s-2-amino-3-hydroxypropanoic acid
PubChem CID 5951
ChEBI CHEBI:17115
IUPAC Name (2S)-2-amino-3-hydroxypropanoic acid
SMILES C(C(C(=O)O)N)O

Specifications

Melting Point 228°C
Color White
Quantity 5 g
Formula Weight 105.09
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Serine
EINECSNumber (9)-1585
RTECSNumber VT8100000
TSCA Yes