CAS RN 3618-43-7
IUPAC Name:
tetrasodium 2-[({5-[3-(3-{[bis(carboxylatomethyl)amino]methyl}-4-hydroxy-5-methylphenyl)-2,2-dioxo-3H-1,2λ⁶-benzoxathiol-3-yl]-2-hydroxy-3-methylphenyl}methyl)(carboxylatomethyl)amino]acetate
Synonyms:
tetrasodium 2-[({5-[3-(3-{[bis(carboxylatomethyl)amino]methyl}-4-hydroxy-5-methylphenyl)-2,2-dioxo-1,2λ⁶-benzoxathiol-3-yl]-2-hydroxy-3-methylphenyl}methyl)(carboxylatomethyl)amino]acetate
Molecular Weight (g/mol):
760.59
Molecular Formula:
C31H28N2Na4O13S
InChi Key:
FXYMNKLSOQONJB-UHFFFAOYSA-J
SMILES:
[Na+].[Na+].[Na+].[Na+].CC1=CC(=CC(CN(CC([O-])=O)CC([O-])=O)=C1O)C1(C2=CC=CC=C2OS1(=O)=O)C1=CC(C)=C(O)C(CN(CC([O-])=O)CC([O-])=O)=C1
1
–
12
of
12
results
1
Thermo Scientific Chemicals Xylenol Orange tetrasodium salt, 0.1% w/v aq. soln.
CAS: 3618-43-7 Molecular Formula: C31H28N2Na4O13S MDL Number: MFCD00044293
2
Thermo Scientific Chemicals Xylenol Orange, sodium salt, pure, water soluble
CAS: 3618-43-7 Molecular Formula: C31H28N2Na4O13S
3
Promotions Available
Thermo Scientific Chemicals Xylenol orange, sodium salt, ACS reagent
Thermo Scientific Chemicals Xylenol orange, sodium salt, ACS reagent
CAS: 3618-43-7 Molecular Formula: C31H28N2Na4O13S Molecular Weight (g/mol): 760.59
4
Thermo Scientific Chemicals Xylenol Orange tetrasodium salt, ACS
CAS: 3618-43-7 MDL Number: MFCD00044293
6
Xylenol Orange Sodium Salt, ACS Grade, LabChem™
CAS: 3618-43-7 Molecular Weight (g/mol): 760.59
7
Xylenol Orange, 0.1% Aqueous, Certified, LabChem™
CAS: 3618-43-7
9
Xylenol Orange Indicator, 0.1% (w/v), Ricca Chemical
CAS: 3618-43-7 Molecular Formula: C31H28N2Na4O13S
10
Xylenol Orange Sodium Salt, ACS Reagent Grade, Ricca Chemical
CAS: 3618-43-7 Molecular Weight (g/mol): 760.58 g/mol
11
12
Xylenol Orange Sodium Salt, ACS Reagent, Reagents
Xylenol Orange Sodium Salt, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.