missing translation for 'onlineSavingsMsg'
Learn More

Xylenol Orange, 0.1% Aqueous, Certified, LabChem™

Catalog No. LC270007
Change view
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC270007 125 mL Poly Bottle
LC270001 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC270007 Supplier LabChem Supplier No. LC270007

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3618-43-7 , 7732-18-5

Specifications

CAS Max % 99.9, 0.1
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1g/mL
Suitability Passes Test
Linear Formula C31H28N2Na4O13S
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide
Density 1g/mL
Color Purple/Red
Solubility Information Soluble in water
Quantity 125 mL
Physical Form Liquid
Molecular Weight (g/mol) 760.59
Formula Weight 760.59
Grade Certified
Chemical Name or Material Xylenol Orange, 0.1% Aqueous
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature