missing translation for 'onlineSavingsMsg'
Learn More

Xylenol Orange Sodium Salt, ACS Reagent, Reagents

Catalog No. 01337628
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337628 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01337628 Supplier Labchem Inc Supplier No. C13483005C2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Xylenol Orange Sodium Salt, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Xylenol Orange Sodium Salt, ACS Reagent
Boiling Point 904°C
CAS 3618-43-7
Packaging Glass Bottle
Specific Gravity 1.0001
Molecular Formula C31H28N2Na4O13S
pH Color Changes Violet/Red to Purple (excess Metal) to Yellow (excess EDTA)
Quantity 5 g
Physical Form Solid
Molecular Weight (g/mol) 760.5837
Grade ACS
Show More Show Less