missing translation for 'onlineSavingsMsg'
Learn More

Xylenol Orange Indicator, 0.2% (w/v) in Alcohol, Ricca Chemical

Catalog No. p-2917284
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
93884 120 mL Natural Poly Bottle
938816 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 93884 Supplier Ricca Chemical Company Supplier No. 93884

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3618-43-7

Specifications

Concentration or Composition (by Analyte or Components) 0.2% (w/v) Alcoholic Solution, in SDA-3A (100 parts Ethanol and 5 parts Methanol)
Packaging Natural Poly Bottle
Quantity 120 mL
Chemical Name or Material Xylenol Orange
ShelfLife 30 days