missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Xylenol orange, sodium salt, ACS reagent

Catalog No. AC447600250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC447600250 25 g Glass bottle
AC447600050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC447600250 Supplier Thermo Scientific Chemicals Supplier No. 447600250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3618-43-7 | C31H28N2Na4O13S | 760.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Chemical Identifiers

CAS 3618-43-7
Molecular Formula C31H28N2Na4O13S
Molecular Weight (g/mol) 760.59

Specifications

Melting Point 210°C
Packaging Glass bottle
Quantity 25 g
Merck Index 15,10281
Solubility Information Solubility in water: 510g/L (20°C)
Formula Weight 760.59
Clarity Passes test
Grade ACS Reagent
Chemical Name or Material Xylenol orange, sodium salt
EINECSNumber 222-805-8

RUO – Research Use Only