missing translation for 'onlineSavingsMsg'
Learn More

Xylenol Orange Indicator, 0.1% (w/v), Ricca Chemical

Catalog No. 93864
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
93864 120 mL Natural Poly Bottle
938616 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 93864 Supplier Ricca Chemical Company Supplier No. 93864

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Indicator for EDTA Titrations

Chemical Identifiers

CAS 3618-43-7 , 7732-18-5
Molecular Formula C31H28N2Na4O13S

Specifications

Packaging Natural Poly Bottle
Color Orange to Dark Purple
Quantity 120 mL
Percent Purity 0.1% W/V
Grade Indicator
Concentration or Composition (by Analyte or Components) 0.1% (w/v) Aqueous Solution
Chemical Name or Material Xylenol Orange
ShelfLife 30 days