CAS RN 29836-26-8
n-Octyl-β-D-glucopyranoside (White Crystalline Powder), Fisher BioReagents
CAS: 29836-26-8 Molecular Formula: C14H28O6 Molecular Weight (g/mol): 292.37 MDL Number: MFCD00063288 InChI Key: HEGSGKPQLMEBJL-RKQHYHRCSA-N PubChem CID: 62852 SMILES: CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O
n-Octyl-beta-D-glucopyranoside
CAS: 29836-26-8 Molecular Formula: C14H28O6 Molecular Weight (g/mol): 292.37 MDL Number: MFCD00063288 InChI Key: HEGSGKPQLMEBJL-RKQHYHRCSA-N Synonym: octyl-beta-d-glucopyranoside,b-octylglucoside,octyl beta-d-glucopyranoside,octyl beta-d-glucoside,octyl glucoside,n-octyl-beta-d-glucoside,octyl b-d-glucopyranoside,2r,3s,4s,5r,6r-2-hydroxymethyl-6-octyloxy tetrahydro-2h-pyran-3,4,5-triol,unii-v109wut6rl,n-octyl glucoside PubChem CID: 62852 IUPAC Name: (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-octoxyoxane-3,4,5-triol SMILES: CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O
n-Octyl-beta-D-glucopyranoside, 98+%
CAS: 29836-26-8 Molecular Formula: C14H28O6 Molecular Weight (g/mol): 292.37 MDL Number: MFCD00063288 InChI Key: HEGSGKPQLMEBJL-RKQHYHRCSA-N Synonym: octyl-beta-d-glucopyranoside,b-octylglucoside,octyl beta-d-glucopyranoside,octyl beta-d-glucoside,octyl glucoside,n-octyl-beta-d-glucoside,octyl b-d-glucopyranoside,2r,3s,4s,5r,6r-2-hydroxymethyl-6-octyloxy tetrahydro-2h-pyran-3,4,5-triol,unii-v109wut6rl,n-octyl glucoside PubChem CID: 62852 SMILES: CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O
MilliporeSigma™ n-Octyl-β-D-glucopyranoside, Calbiochem™,
CAS: 29836-26-8 Molecular Formula: C14H28O6 Molecular Weight (g/mol): 292.37 MDL Number: MFCD00063288 InChI Key: HEGSGKPQLMEBJL-RKQHYHRCSA-N Synonym: OG, n-Octylglucoside PubChem CID: 62852 IUPAC Name: (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-(octyloxy)oxane-3,4,5-triol SMILES: CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O
MilliporeSigma™ n-Octyl-β-D-glucopyranoside, ULTROL™ Grade, Calbiochem™,
CAS: 29836-26-8 Molecular Formula: C14H28O6 Molecular Weight (g/mol): 292.37 MDL Number: MFCD00063288 InChI Key: HEGSGKPQLMEBJL-RKQHYHRCSA-N Synonym: OG, n-Octylglucoside PubChem CID: 62852 IUPAC Name: (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-(octyloxy)oxane-3,4,5-triol SMILES: CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O
1-O-n-Octyl-beta-D-glucopyranoside, 98%
CAS: 29836-26-8 Molecular Formula: C14H28O6 Molecular Weight (g/mol): 292.37 MDL Number: MFCD00063288 InChI Key: HEGSGKPQLMEBJL-RKQHYHRCSA-N Synonym: octyl-beta-d-glucopyranoside,b-octylglucoside,octyl beta-d-glucopyranoside,octyl beta-d-glucoside,octyl glucoside,n-octyl-beta-d-glucoside,octyl b-d-glucopyranoside,2r,3s,4s,5r,6r-2-hydroxymethyl-6-octyloxy tetrahydro-2h-pyran-3,4,5-triol,unii-v109wut6rl,n-octyl glucoside PubChem CID: 62852 IUPAC Name: (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-octoxyoxane-3,4,5-triol SMILES: CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O
n-Octyl beta-D-Glucopyranoside 96.0+%, TCI America™
CAS: 29836-26-8 Molecular Formula: C14H28O6 Molecular Weight (g/mol): 292.37 MDL Number: MFCD00063288 InChI Key: HEGSGKPQLMEBJL-RKQHYHRCSA-N Synonym: octyl-beta-d-glucopyranoside,b-octylglucoside,octyl beta-d-glucopyranoside,octyl beta-d-glucoside,octyl glucoside,n-octyl-beta-d-glucoside,octyl b-d-glucopyranoside,2r,3s,4s,5r,6r-2-hydroxymethyl-6-octyloxy tetrahydro-2h-pyran-3,4,5-triol,unii-v109wut6rl,n-octyl glucoside PubChem CID: 62852 IUPAC Name: (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-(octyloxy)oxane-3,4,5-triol SMILES: CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O
n-Octyl beta-D-Glucopyranoside 96.0+%, TCI America™
CAS: 29836-26-8 Molecular Formula: C14H28O6 Molecular Weight (g/mol): 292.37 MDL Number: MFCD00063288 InChI Key: HEGSGKPQLMEBJL-RKQHYHRCSA-N Synonym: octyl-beta-d-glucopyranoside,b-octylglucoside,octyl beta-d-glucopyranoside,octyl beta-d-glucoside,octyl glucoside,n-octyl-beta-d-glucoside,octyl b-d-glucopyranoside,2r,3s,4s,5r,6r-2-hydroxymethyl-6-octyloxy tetrahydro-2h-pyran-3,4,5-triol,unii-v109wut6rl,n-octyl glucoside PubChem CID: 62852 IUPAC Name: (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-(octyloxy)oxane-3,4,5-triol SMILES: CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O
n-Octyl-β-D-Glucopyranoside, MP Biomedicals™
CAS: 29836-26-8 Molecular Formula: C14H28O6 Molecular Weight (g/mol): 292.37 MDL Number: MFCD00063288 InChI Key: HEGSGKPQLMEBJL-RKQHYHRCSA-N Synonym: n-Octyl-b-D-glucoside,OGP PubChem CID: 62852 SMILES: CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O