missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Octyl beta-D-Glucopyranoside 96.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 5 g |
| Formula Weight | 292.37 |
| Percent Purity | ≥96.0% (GC) |
| Grade | Not Available |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | n-Octyl beta-D-Glucopyranoside |
Safety and Handling
| EINECSNumber | (5)-6337&(5)-3641 |
| TSCA | No |