missing translation for 'onlineSavingsMsg'
Learn More

n-Octyl-beta-D-glucopyranoside

Catalog No. AAL13259MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL13259MD 250 mg
AAL1325903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL13259MD Supplier Thermo Scientific Chemicals Supplier No. L13259MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 29836-26-8 | C14H28O6 | 292.37 g/mol

2-Bromo-4'-(diethylamino)acetophenone, is used as non-ionic dialyzable detergent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-4′-(diethylamino)acetophenone, is used as non-ionic dialyzable detergent.

Solubility
Insoluble in water.

Notes
Heat and moisture sensitive. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Chemical Identifiers

CAS 29836-26-8
Molecular Formula C14H28O6
Molecular Weight (g/mol) 292.37
MDL Number MFCD00063288
InChI Key HEGSGKPQLMEBJL-RKQHYHRCSA-N
Synonym octyl-beta-d-glucopyranoside, b-octylglucoside, octyl beta-d-glucopyranoside, octyl beta-d-glucoside, octyl glucoside, n-octyl-beta-d-glucoside, octyl b-d-glucopyranoside, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-octyloxy tetrahydro-2h-pyran-3,4,5-triol, unii-v109wut6rl, n-octyl glucoside
PubChem CID 62852
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-octoxyoxane-3,4,5-triol
SMILES CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O

Specifications

Melting Point 103°C to 104°C
Quantity 250 mg
Beilstein 84118
Solubility Information Insoluble in water.
Optical Rotation −31° (c=2 in Methanol)
Formula Weight 292.37
Chemical Name or Material n-Octyl-beta-D-glucopyranoside
EINECSNumber 249-887-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only